Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Isopropyl-3-methylphenol 99.0+%, TCI America™
SDP

Catalog No. M0409100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0409100G 100 g
M040925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M0409100G Supplier TCI America Supplier No. M0409100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3228-02-2
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.22
MDL Number MFCD00010704
InChI Key IJALWSVNUBBQRA-UHFFFAOYSA-N
Synonym Biosol, 4-Isopropyl-m-cresol
PubChem CID 18597
ChEBI CHEBI:34429
IUPAC Name 3-methyl-4-propan-2-ylphenol
SMILES CC1=C(C=CC(=C1)O)C(C)C

Specifications

Melting Point 112°C
Color White-Yellow
Boiling Point 246°C
Quantity 100 g
Formula Weight 150.22
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Isopropyl-3-methylphenol
EINECSNumber (3)-0521&(4)-0057
RTECSNumber GZ7170000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.