Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Isopropylamino)butanol 98.0+%, TCI America™
SDP

Catalog No. I07561G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I07561G 1 g
I07565G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I07561G Supplier TCI America Supplier No. I07561G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 42042-71-7
Molecular Formula C7H17NO
Molecular Weight (g/mol) 131.219
InChI Key IPLWOCGPIGUXOR-UHFFFAOYSA-N
PubChem CID 12950986
IUPAC Name 4-(propan-2-ylamino)butan-1-ol
SMILES CC(C)NCCCCO

Specifications

Color Colorless
Boiling Point 85°C
Quantity 1 g
UN Number 1993
Formula Weight 131.22
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-(Isopropylamino)butanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.