Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Isopropylbenzyl alcohol, 97%

Catalog No. p-3778901
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle

CAS: 536-60-7 | C10H14O | 150.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 536-60-7
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.22
MDL Number MFCD00004663
InChI Key OIGWAXDAPKFNCQ-UHFFFAOYSA-N
Synonym 4-isopropylbenzyl alcohol, cuminol, cumic alcohol, cuminyl alcohol, cuminic alcohol, 4-isopropylphenyl methanol, cumyl alcohol, p-cymen-7-ol, cumin alcohol, p-isopropylbenzyl alcohol
PubChem CID 325
ChEBI CHEBI:27628
IUPAC Name (4-propan-2-ylphenyl)methanol
SMILES CC(C)C1=CC=C(C=C1)CO

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 28°C
Color Colorless to Yellow
Density 0.9800g/mL
Boiling Point 248°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5180 to 1.52
Linear Formula (CH3)2CHC6H4CH2OH
Quantity 25 g
Beilstein 06, III, 1911
Specific Gravity 0.98
Merck Index 15, 2609
Formula Weight 150.22
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4-Isopropylbenzyl alcohol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face,hands and any exposed skin thoroughly after handling.
EINECSNumber 208-640-4

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.