Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Isopropylbenzyl Alcohol 98.0+%, TCI America™
SDP

Catalog No. I0272100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0272100ML 100 mL
I027225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I0272100ML Supplier TCI America Supplier No. I0272100ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 536-60-7
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.221
MDL Number MFCD00004663
InChI Key OIGWAXDAPKFNCQ-UHFFFAOYSA-N
Synonym 4-isopropylbenzyl alcohol, cuminol, cumic alcohol, cuminyl alcohol, cuminic alcohol, 4-isopropylphenyl methanol, cumyl alcohol, p-cymen-7-ol, cumin alcohol, p-isopropylbenzyl alcohol
PubChem CID 325
ChEBI CHEBI:27628
IUPAC Name (4-propan-2-ylphenyl)methanol
SMILES CC(C)C1=CC=C(C=C1)CO

Specifications

Color Yellow
Boiling Point 248°C
Quantity 100 mL
Formula Weight 150.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Isopropylbenzyl Alcohol
EINECSNumber (3)-1049
RTECSNumber GZ7260000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.