Alfa Aesar™ 4-Isopropylphenol, 98%

Manufacturer: Alfa Aesar™ B22620A1

Quantity 1kg

 View more versions of this product

Catalog No. AAB22620A1

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 99-89-8
    Molecular Formula C9H12O
    Formula Weight 136.2
    MDL Number MFCD00002372
    UN Number UN2430
    Beilstein 1363564
    Synonym p-Cumenol; p-Hydroxycumene
    Chemical Name or Material 4-Isopropylphenol

    Specifications

    Formula Weight 136.2
    Molecular Formula C9H12O
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    RTECS Number SL5950000
    Flash Point 108°C (226°F)
    Boiling Point 211° to 212°C
    Melting Point 59° to 61°C
    TSCA TSCA
    MDL Number MFCD00002372
    CAS 99-89-8
    UN Number UN2430
    Beilstein 1363564
    Synonym p-Cumenol; p-Hydroxycumene
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318-H302
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ALKYLPHENOLS, SOLID, N.O.S.
    Quantity 1kg
    Chemical Name or Material 4-Isopropylphenol
    Assay 98%

    Description

    Applications
    4-Isopropylphenol was used in the synthesis of vinyl compounds. It is used as a flavor & fragrance agent in cosmetics.

    Solubility
    Slightly soluble in water, soluble in ethanol and ethyl ether.

    Notes
    Incompatible with oxidizing agents.

    GHS H Statement
    H314-H318-H302
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit