Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Isopropylphenol, 98%, Thermo Scientific Chemicals

Catalog Number AAB2262018
Click to view available options
Quantity:
50 g
250 g
1 kg
99-89-8
C9H12O
136.194
MFCD00002372
YQUQWHNMBPIWGK-UHFFFAOYSA-N
4-isopropylphenol, p-isopropylphenol, p-cumenol, australol, phenol, 4-1-methylethyl, 4-1-methylethyl phenol, 1-hydroxy-4-isopropylbenzene, 4-propan-2-yl phenol, phenol, p-isopropyl, para-isopropylphenol
7465
4-propan-2-ylphenol
CC(C)C1=CC=C(C=C1)O
99-89-8
C9H12O
136.194
MFCD00002372
YQUQWHNMBPIWGK-UHFFFAOYSA-N
4-isopropylphenol, p-isopropylphenol, p-cumenol, australol, phenol, 4-1-methylethyl, 4-1-methylethyl phenol, 1-hydroxy-4-isopropylbenzene, 4-propan-2-yl phenol, phenol, p-isopropyl, para-isopropylphenol
7465
4-propan-2-ylphenol
CC(C)C1=CC=C(C=C1)O

4-Isopropylphenol was used in the synthesis of vinyl compounds. It is used as a flavor & fragrance agent in cosmetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Isopropylphenol was used in the synthesis of vinyl compounds. It is used as a flavor & fragrance agent in cosmetics.

Solubility
Slightly soluble in water, soluble in ethanol and ethyl ether.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 59°C to 61°C
Boiling Point 211°C to 212°C
Flash Point 108°C (226°F)
Quantity 50 g
UN Number UN2430
Beilstein 1363564
Solubility Information Slightly soluble in water,soluble in ethanol and ethyl ether.
Formula Weight 136.2
Percent Purity 98%
Chemical Name or Material 4-Isopropylphenol
Hazard Category H302+H312+H332-H314-H334-H335
Hazard Statement GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P260-P264b-P270-P271-P280-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ALKYLPHENOLS, SOLID, N.O.S.
EINECSNumber 202-798-8
RTECSNumber SL5950000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit