Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Ketopimelic acid, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
502-50-1
C7H10O5
174.15
MFCD00002798
UDDSEESQRGPVIL-UHFFFAOYSA-N
4-ketopimelic acid,4-oxopimelic acid,4-ketopimelic,.gamma.-ketopimelic acid,heptanedioic acid, 4-oxo,acmc-20amfs,pimelic acid, 4-keto,heptanedioic acid,4-oxo,4-oxoheptanedioic acid
95084
4-oxoheptanedioic acid
C(CC(=O)O)C(=O)CCC(=O)O
502-50-1
C7H10O5
174.15
MFCD00002798
UDDSEESQRGPVIL-UHFFFAOYSA-N
4-ketopimelic acid,4-oxopimelic acid,4-ketopimelic,.gamma.-ketopimelic acid,heptanedioic acid, 4-oxo,acmc-20amfs,pimelic acid, 4-keto,heptanedioic acid,4-oxo,4-oxoheptanedioic acid
95084
4-oxoheptanedioic acid
C(CC(=O)O)C(=O)CCC(=O)O
Infrared Spectrum Authentic
Color White to Yellow
Melting Point 142.0°C to 144.0°C
Packaging Glass bottle
Quantity 100g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula HO2CCH2CH2COCH2CH2CO2H
Beilstein 03, 804
Formula Weight 174.15
Physical Form Powder, Crystals and/or Chunks
Percent Purity 98%
Chemical Name or Material 4-Ketopimelic acid
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning