Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Maleimidobutyric Acid 98.0+%, TCI America™
SDP

Catalog No. M23371G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M23371G 1 g
M23375G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M23371G Supplier TCI America Supplier No. M23371G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 57078-98-5
Molecular Formula C8H9NO4
Molecular Weight (g/mol) 183.163
MDL Number MFCD00043139
InChI Key NCPQROHLJFARLL-UHFFFAOYSA-N
PubChem CID 3404904
IUPAC Name 4-(2,5-dioxopyrrol-1-yl)butanoic acid
SMILES C1=CC(=O)N(C1=O)CCCC(=O)O

Specifications

Melting Point 96°C
Color White-Yellow
Quantity 1 g
Formula Weight 183.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Maleimidobutyric Acid
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.