Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-Mercaptobenzoic acid

Catalog No. 6625345G Shop All MilliporeSigma Sigma Organic Chemistry Products
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
66-253-45G 5 g
66-253-425G 25 g
2 options

Catalog No. 66-253-45G

Supplier: Sigma Aldrich 6625345G

Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

4-Mercaptobenzoic acid

Specifications

CAS 1074-36-8
Melting Point 215°C to 224°C (lit.)
Molecular Formula C7H6O2S
Linear Formula HSC6H4CO2H
MDL Number MFCD00016617
Quantity 5 g
Molecular Weight (g/mol) 154.19
Percent Purity 90%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.