Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Mercaptobenzoic Acid 95.0+%, TCI America™
SDP

Catalog No. m12945g
Click to view available options
Quantity:
5 g

Chemical Identifiers

CAS 1074-36-8
Molecular Formula C7H6O2S
Molecular Weight (g/mol) 154.18
MDL Number MFCD00016617
InChI Key LMJXSOYPAOSIPZ-UHFFFAOYSA-N
Synonym 4-mercaptobenzoic acid, 4-mercaptobenzoate, benzoic acid, 4-mercapto, 4-mercapto-benzoic acid, 4-mercapto benzoic acid, 4-thiobenzoic acid, p-mercaptobenzoic acid, benzoic acid, p-mercapto, para-mercaptobenzoate, paracarboxythiophenol
PubChem CID 95738
IUPAC Name 4-sulfanylbenzoic acid
SMILES OC(=O)C1=CC=C(S)C=C1

Specifications

Melting Point 219°C
Quantity 5 g
Formula Weight 154.18
Percent Purity ≥95.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Mercaptobenzoic Acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.