Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxy-3-(trifluoromethyl)benzoic Acid 98.0+%, TCI America™
SDP

Catalog No. M309525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M309525G 25 g
M30955G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M309525G Supplier TCI America Supplier No. M309525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 213598-09-5
Molecular Formula C9H7F3O3
Molecular Weight (g/mol) 220.15
MDL Number MFCD01091014
InChI Key VNWUPESTVBTYJM-UHFFFAOYSA-N
Synonym 4-methoxy-3-trifluoromethyl benzoic acid, 4-methoxy-3-trifluoromethyl-benzoic acid, acmc-1cr7h, 3-trifluoromethyl-4-methoxybenzoic acid, benzoic acid, 4-methoxy-3-trifluoromethyl, 4-methyloxy-3-trifluoromethyl benzoic acid
PubChem CID 2775295
IUPAC Name 4-methoxy-3-(trifluoromethyl)benzoic acid
SMILES COC1=CC=C(C=C1C(F)(F)F)C(O)=O

Specifications

Melting Point 172°C
Quantity 25 g
Formula Weight 220.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxy-3-(trifluoromethyl)benzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.