Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxy-4'-nitrobenzophenone, 97%

Catalog No. p-7026624
Click to view available options
Quantity:
1 g
5 g
25 g

CAS: 1151-94-6 | C14H11NO4 | 257.25 g/mol

4-Methoxy-4'-nitrobenzophenone is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxy-4′-nitrobenzophenone is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1151-94-6
Molecular Formula C14H11NO4
Molecular Weight (g/mol) 257.25
MDL Number MFCD00178348
InChI Key DXVSAELNVPXMSQ-UHFFFAOYSA-N
Synonym 4-methoxy-4'-nitrobenzophenone, 4-methoxyphenyl 4-nitrophenyl methanone, 4-methoxyphenyl-4-nitrophenyl methanone, methanone, 4-methoxyphenyl 4-nitrophenyl, 4-methoxy-phenyl-4-nitro-phenyl-methanone, acmc-1buw2, 4-methoxy-4-nitrobenzophenone, 4-methoxy-4?-nitrobenzophenone, 4-methoxy-4/'-nitrobenzophenone
PubChem CID 347772
IUPAC Name (4-methoxyphenyl)-(4-nitrophenyl)methanone
SMILES COC1=CC=C(C=C1)C(=O)C1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 125°C to 127°C
Quantity 1 g
Formula Weight 257.25
Percent Purity 97%
Chemical Name or Material 4-Methoxy-4'-nitrobenzophenone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.