Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxy-N-(2-methylbenzyl)benzamide, 97%, Thermo Scientific™

Catalog No. AAH5934603
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5934603 1 g
AAH59346MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5934603 Supplier Thermo Scientific Chemicals Supplier No. H5934603

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 331638-66-5
Molecular Formula C16H17NO2
Molecular Weight (g/mol) 255.317
MDL Number MFCD01154205
InChI Key CVDCGDPYWBZBAN-UHFFFAOYSA-N
Synonym 4-methoxy-n-2-methylbenzyl benzamide, 4-methoxy-n-2-methylphenyl methyl benzamide, cbmicro_011456, cambridge id 6073509, 4-hydroxymethyl-n-2-methylbenzyl benzamide
PubChem CID 789734
IUPAC Name 4-methoxy-N-[(2-methylphenyl)methyl]benzamide
SMILES CC1=CC=CC=C1CNC(=O)C2=CC=C(C=C2)OC

Specifications

Quantity 1 g
Formula Weight 274.31
Percent Purity 97%
Chemical Name or Material 4-Methoxy-N-(2-methylbenzyl)benzamide
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.