Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Methoxyacetophenone, 99%

Catalog No. AAA1116230
Click to view available options
Quantity:
25 g
250 g
1000 g

CAS: 100-06-1 | C9H10O2 | 150.18 g/mol

4'-Methoxyacetophenone is used as a component of perfumes and as chemical intermediate in the manufacture of pharmaceuticals, resins, flavouring agents. And also used to study ruthenium catalyzed step growth copolymerization of 4?-methoxyacetophenone with α,ω-dienes to yield copolymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′-Methoxyacetophenone is used as a component of perfumes and as chemical intermediate in the manufacture of pharmaceuticals, resins, flavouring agents. And also used to study ruthenium catalyzed step growth copolymerization of 4′-methoxyacetophenone with α,ω-dienes to yield copolymers.

Solubility
Insoluble in water.

Notes
Light Sensitive, Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-06-1
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.18
MDL Number MFCD00008745
InChI Key NTPLXRHDUXRPNE-UHFFFAOYSA-N
Synonym 4'-methoxyacetophenone, 4-acetylanisole, 4-methoxyacetophenone, 1-4-methoxyphenyl ethanone, p-methoxyacetophenone, acetanisole, novatone, linarodin, vananote, ethanone, 1-4-methoxyphenyl
PubChem CID 7476
ChEBI CHEBI:86567
IUPAC Name 1-(4-methoxyphenyl)ethanone
SMILES COC1=CC=C(C=C1)C(C)=O

Specifications

Melting Point 36°C to 40°C
Density 1.08
Boiling Point 258°C to 260°C
Flash Point 138°C (280°F)
Odor Characteristic
Quantity 250 g
Beilstein 742313
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 150.18
Percent Purity 99%
Chemical Name or Material 4'-Methoxyacetophenone
Show More Show Less
Hazard Category H302
Hazard Statement GHS H Statement
H301-H315
Toxic if swallowed.
Causes skin irritation.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 202-815-9
RTECSNumber AM9240000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.