Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxybenzamide, 97%

Catalog No. p-156917
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle

CAS: 3424-93-9 | C8H9NO2 | 151.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3424-93-9
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.16
MDL Number MFCD00007995
InChI Key GUCPYIYFQVTFSI-UHFFFAOYSA-N
Synonym p-anisamide, p-methoxybenzamide, benzamide, 4-methoxy, 4-methoxy benzamide, p-methyoxybenzamide, unii-z85fx3977e, 4-methoxy-benzamide, 4-anisamide, acmc-1cjjo, 4-methoxylbenzamide
PubChem CID 76959
IUPAC Name 4-methoxybenzamide
SMILES COC1=CC=C(C=C1)C(=O)N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 164.0°C to 167.0°C
Color White
Boiling Point 295.0°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula CH3OC6H4CONH2
Quantity 5 g
Beilstein 10, II, 100
Solubility Information Solubility in water: soluble
Formula Weight 151.16
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxybenzamide, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 222-319-6
RTECSNumber CV5466666

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.