Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxybenzenesulfonamide, 98%

Catalog No. aal0896106
Click to view available options
Quantity:
5 g
25 g

CAS: 1129-26-6 | C7H9NO3S | 187.213 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1129-26-6
Molecular Formula C7H9NO3S
Molecular Weight (g/mol) 187.213
MDL Number MFCD00025392
InChI Key MSFQEZBRFPAFEX-UHFFFAOYSA-N
Synonym 4-methoxybenzenesulphonamide, p-methoxybenzenesulfonamide, 4-methoxybenzene-1-sulfonamide, benzenesulfonamide, 4-methoxy, 4-methoxy-benzenesulfonamide, benzenesulfonamide, p-methoxy, benzenesulfonamide, p-methoxy-6ci,7ci,8ci, 3lka, p-anisolesulfonamide, acmc-1bvgf
PubChem CID 70789
IUPAC Name 4-methoxybenzenesulfonamide
SMILES COC1=CC=C(C=C1)S(=O)(=O)N

Specifications

Melting Point 110°C to 114°C
Quantity 5 g
Beilstein 2047881
Solubility Information Slightly soluble in water.
Formula Weight 187.22
Percent Purity 98%
Chemical Name or Material 4-Methoxybenzenesulfonamide
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
RTECSNumber DB2900000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.