Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxybenzyl 2,2,2-Trichloroacetimidate 96.0+%, TCI America™
SDP

Catalog No. M201625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M201625G 25 g
M20165G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M201625G Supplier TCI America Supplier No. M201625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 89238-99-3
Molecular Formula C10H10Cl3NO2
Molecular Weight (g/mol) 282.545
MDL Number MFCD00134547
InChI Key TYHGKLBJBHACOI-UHFFFAOYSA-N
Synonym 2,2,2-Trichloroacetimidic Acid 4-Methoxybenzyl Ester
PubChem CID 11087263
IUPAC Name (4-methoxyphenyl)methyl 2,2,2-trichloroethanimidate
SMILES COC1=CC=C(C=C1)COC(=N)C(Cl)(Cl)Cl

Specifications

Color Yellow
Boiling Point 137°C
Quantity 25 g
Formula Weight 282.55
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Methoxybenzyl 2,2,2-Trichloroacetimidate
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.