Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Methoxychalcone, 97%

Catalog No. AAB2215814
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 959-23-9 | C16H14O2 | 238.286 g/mol

4-Methoxychalcone is used as a precursor to prepare 3-(4-methoxy-phenyl)-3-morpholin-4-yl-1-phenyl-propan-1-one by reacting with morpholine in presence of heptanes as solvent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxychalcone is used as a precursor to prepare 3-(4-methoxy-phenyl)-3-morpholin-4-yl-1-phenyl-propan-1-one by reacting with morpholine in presence of heptanes as solvent.

Solubility
Soluble in dichloromethane and methanol. Insoluble in water.

Notes
Keep the container tightly closed in a dry and well-ventilated place. Incompatible withstrong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 959-23-9
Molecular Formula C16H14O2
Molecular Weight (g/mol) 238.286
MDL Number MFCD00008407
InChI Key KJHHAPASNNVTSN-KPKJPENVSA-N
Synonym 4'-methoxychalcone, 2e-1-4-methoxyphenyl-3-phenylprop-2-en-1-one, 1-4-methoxyphenyl-3-phenylprop-2-en-1-one, chembl34398, 1-4-methoxyphenyl-3-phenyl-2-propen-1-one, e-1-4-methoxyphenyl-3-phenylprop-2-en-1-one, unii-bm45n45fiz, e-1-4-methoxy-phenyl-3-phenyl-propenone, 2-propen-1-one, 1-4-methoxyphenyl-3-phenyl-, 2e, 2-propen-1-one, 1-4-methoxyphenyl-3-phenyl
PubChem CID 641818
IUPAC Name (E)-1-(4-methoxyphenyl)-3-phenylprop-2-en-1-one
SMILES COC1=CC=C(C=C1)C(=O)C=CC2=CC=CC=C2

Specifications

Melting Point 101°C to 103°C
Quantity 25 g
Beilstein 644640
Solubility Information Soluble in dichloromethane and methanol. Insoluble in water.
Formula Weight 238.29
Percent Purity 97%
Chemical Name or Material 4'-Methoxychalcone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 213-495-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.