Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxyphenol, 98+%

Catalog No. aaa1484518
Click to view available options
Quantity:
50 g
250 g
1000 g

CAS: 150-76-5 | C7H8O2 | 124.14 g/mol

4-Methoxyphenol is an active ingredient and used in dermatology. It is employed as a pharmaceutical drug in skin depigmentation. It is used as polymerization inhibitors. For example, in the radical polymerization of acryaltes and styrene monomers. It is also used as an intermediate in the preparationagrochemicals, liquid crystals. It acts as a stabilizer for the formulation of inks, toners and adhesives. It is mainly used as an additive for textile and leather industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxyphenol is an active ingredient and used in dermatology. It is employed as a pharmaceutical drug in skin depigmentation. It is used as polymerization inhibitors. For example, in the radical polymerization of acryaltes and styrene monomers. It is also used as an intermediate in the preparationagrochemicals, liquid crystals. It acts as a stabilizer for the formulation of inks, toners and adhesives. It is mainly used as an additive for textile and leather industries.

Solubility
Soluble in acetone, ethyl acetate, ethanol, ether, benzene and carbon tetrachloride.

Notes
Moisture sensitive. Store in a cool place. Keep the container tightly closed. Incompatible with halogens and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150-76-5
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.14
MDL Number MFCD00002332
InChI Key NWVVVBRKAWDGAB-UHFFFAOYSA-N
Synonym mequinol, 4-hydroxyanisole, p-hydroxyanisole, p-methoxyphenol, hydroquinone monomethyl ether, phenol, 4-methoxy, leucobasal, mehq, leucodine b, mechinolum
PubChem CID 9015
ChEBI CHEBI:69441
IUPAC Name 4-methoxyphenol
SMILES COC1=CC=C(O)C=C1

Specifications

Melting Point 55°C to 58°C
Density 1.55
Boiling Point 243°C to 246°C
Flash Point 132°C (269°F)
Quantity 50 g
Beilstein 507924
Solubility Information Soluble in acetone,ethyl acetate,ethanol,ether,benzene and carbon tetrachloride.
Formula Weight 124.14
Percent Purity ≥98%
Chemical Name or Material 4-Methoxyphenol
Hazard Category H302-H317-H319-H361d-H373-H500
Hazard Statement GHS H Statement
H302-H319-H317
Harmful if swallowed.
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P260-P264b-P270-P272-P280-P281-P301+P312-P302+P352-P305+P351+P338-P308+P313-P330-P333+P313-P363-P501c
EINECSNumber 205-769-8
RTECSNumber SL7700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.