Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxyphenyl 4-Hydroxybenzoate 99.0+%, TCI America™
SDP

Catalog No. H082525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H082525G 25 g
H08255G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H082525G Supplier TCI America Supplier No. H082525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 50687-62-2
Molecular Formula C14H12O4
Molecular Weight (g/mol) 244.246
MDL Number MFCD00191515
InChI Key MTQSVJUCLQYISS-UHFFFAOYSA-N
Synonym 4-Hydroxybenzoic Acid 4-Methoxyphenyl Ester, 4-Methoxyphenylparaben
PubChem CID 578384
IUPAC Name (4-methoxyphenyl) 4-hydroxybenzoate
SMILES COC1=CC=C(C=C1)OC(=O)C2=CC=C(C=C2)O

Specifications

Melting Point 194°C
Color White
Quantity 25 g
Formula Weight 244.25
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxyphenyl 4-Hydroxybenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.