Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(4-Methoxyphenyl)acetonitrile, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
104-47-2
C9H9NO
147.177
MFCD00001919
PACGLQCRGWFBJH-UHFFFAOYSA-N
4-methoxyphenylacetonitrile,2-4-methoxyphenyl acetonitrile,4-methoxybenzyl cyanide,4-methoxyphenyl acetonitrile,p-methoxyphenylacetonitrile,benzeneacetonitrile, 4-methoxy,p-methoxybenzyl cyanide,anisylacetonitrile,4-anisyl cyanide,p-methoxybenzeneacetonitrile
66031
2-(4-methoxyphenyl)acetonitrile
COC1=CC=C(C=C1)CC#N
104-47-2
C9H9NO
147.177
MFCD00001919
PACGLQCRGWFBJH-UHFFFAOYSA-N
4-methoxyphenylacetonitrile,2-4-methoxyphenyl acetonitrile,4-methoxybenzyl cyanide,4-methoxyphenyl acetonitrile,p-methoxyphenylacetonitrile,benzeneacetonitrile, 4-methoxy,p-methoxybenzyl cyanide,anisylacetonitrile,4-anisyl cyanide,p-methoxybenzeneacetonitrile
66031
2-(4-methoxyphenyl)acetonitrile
COC1=CC=C(C=C1)CC#N
Refractive Index 1.53
Color Colorless to Yellow
Infrared Spectrum Authentic
Melting Point 8.0°C
Flash Point 117°C
Density 1.0800g/mL
Boiling Point 286.0°C to 287.0°C
Quantity 100g
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula CH3OC6H4CH2CN
Beilstein 10, 191
Formula Weight 147.18
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material (4-Methoxyphenyl)acetonitrile
Show More Show Less

GHS H Statement:
Toxic if swallowed.
Harmful to aquatic life with long lasting effects.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Rinse mouth.
Avoid release to the environment.
Wash face, hands and any exposed skin thoroughly after handling.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger