Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxyphenylacetic Acid 99%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
250g
1kg
Packaging:
Glass bottle
Plastic bottle
104-01-8
C9H10O3
166.18
MFCD00004345
NRPFNQUDKRYCNX-UHFFFAOYSA-N
4-methoxyphenylacetic acid,homoanisic acid,2-4-methoxyphenyl acetic acid,4-methoxybenzeneacetic acid,benzeneacetic acid, 4-methoxy,4-methoxyphenyl acetic acid,p-methoxyphenylacetic acid,2-p-anisyl acetic acid,p-methoxyphenyl acetic acid
7690
CHEBI:55501
2-(4-methoxyphenyl)acetic acid
COC1=CC=C(C=C1)CC(=O)O
104-01-8
C9H10O3
166.18
MFCD00004345
NRPFNQUDKRYCNX-UHFFFAOYSA-N
4-methoxyphenylacetic acid,homoanisic acid,2-4-methoxyphenyl acetic acid,4-methoxybenzeneacetic acid,benzeneacetic acid, 4-methoxy,4-methoxyphenyl acetic acid,p-methoxyphenylacetic acid,2-p-anisyl acetic acid,p-methoxyphenyl acetic acid
7690
CHEBI:55501
2-(4-methoxyphenyl)acetic acid
COC1=CC=C(C=C1)CC(=O)O
Color White to Yellow
Boiling Point 140.0°C (3.0 mmHg)
Quantity 250g
Melting Point 84.0°C to 88.0°C
Packaging Plastic bottle
Infrared Spectrum Authentic
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula CH3OC6H4CH2CO2H
Beilstein 10, 190
Solubility Information Solubility in water: 6g/L (20 c)
Formula Weight 166.18
Physical Form Crystalline Powder and Chunks
Percent Purity 99%
Chemical Name or Material 4-Methoxyphenylacetic acid, 99%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 203-166-4

RTECSNumber : AI8960000

TSCA : TSCA