Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxyphenylmagnesium bromide, 1M solution in THF, AcroSeal™

Catalog No. p-2799215
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 13139-86-1 | C7H7BrMgO | 211.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13139-86-1, 109-99-9
Molecular Formula C7H7BrMgO
Molecular Weight (g/mol) 211.34
MDL Number MFCD00013515
InChI Key JKXAZEXIOPFJSC-UHFFFAOYSA-M
Synonym 4-methoxyphenylmagnesium bromide, 4-methoxyphenylmagnesium bromide solution, bromo 4-methoxyphenyl magnesium, p-methoxyphenylmagnesium bromide, 4-methoxyphenyl magnesium bromide, 4-methoxyphenymagnesium bromide, 4-methoxy phenylmagnesiumbromide, p-methoxyphenyl magnesium bromide, p-methoxyphenyl-magnesium bromide, 4-methoxy-phenyl-magnesiumbromide, grignard reagent
PubChem CID 2734898
IUPAC Name magnesium;methoxybenzene;bromide
SMILES COC1=CC=C([Mg]Br)C=C1

Specifications

CAS Min % 19.02, 74.64
CAS Max % 25.36, 80.98
Color Yellow to Brown
Density 1.0000g/mL
Flash Point −21°C
Packaging AcroSeal™ Glass bottle
Linear Formula CH3O(C6H4)MgBr
Quantity 100 mL
Specific Gravity 1
Solubility Information Solubility in water: vigorous reaction
Formula Weight 211.34
Physical Form Liquid
Chemical Name or Material 4-Methoxyphenylmagnesium bromide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Highly flammable liquid and vapor.
May form explosive peroxides.
Reacts violently with water.
Suspected of causing cancer.<br/
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p
Recommended Storage Magnesium salts may crystallize below 20°C

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.