Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Methoxypropiophenone, 99%

Catalog No. p-158597
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle

CAS: 121-97-1 | C10H12O2 | 164.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-97-1
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.2
MDL Number MFCD00009310
InChI Key ZJVAWPKTWVFKHG-UHFFFAOYSA-N
PubChem CID 67144
IUPAC Name 1-(4-methoxyphenyl)propan-1-one
SMILES CCC(=O)C1=CC=C(C=C1)OC

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 27°C to 29°C
Color Amber to Colorless
Density 1.0700g/mL
Boiling Point 273°C to 275°C
Flash Point 61°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.5455 to 1.5475
Linear Formula CH3OC6H4COC2H5
Quantity 5 g
Beilstein 08, 103
Specific Gravity 1.07
Synonym 4'-methoxypropiophenone, p-methoxypropiophenone, 4-methoxypropiophenone, 1-4-methoxyphenyl propan-1-one, 1-propanone, 1-4-methoxyphenyl, propiophenone, 4'-methoxy, ethyl 4-methoxyphenyl ketone, 1-4-methoxyphenyl-1-propanone, 1-propanone,1-4-methoxyphenyl, 1-4-methoxy-phenyl-propan-1-one
Formula Weight 164.2
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 4'-Methoxypropiophenone, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 204-512-7
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.