Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium bromide, 4M solution in THF, AcroSeal™

Catalog No. AC390978000
Click to view available options
Quantity:
800 mL
Packaging:
AcroSeal™ Glass Bottle

CAS: 7550-35-8 | BrLi | 86.84 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Lithium bromide
CAS 7550-35-8, 109-99-9
Quantity 800 mL
Molecular Formula BrLi
Linear Formula LiBr
MDL Number MFCD00011077
Flash Point −22°C
Merck Index 15, 5582
Synonym lithium bromide, lithium monobromide, lithium bromide libr, lithiumbromide, libr, lithium 1+ ion bromide, lithium bromide, anhydrous, lithium bromide, ultra dry, bromolithium, lithium-bromide
InChI Key AMXOYNBUYSYVKV-UHFFFAOYSA-M
SMILES [Li+].[Br-]
IUPAC Name lithium;bromide
Molecular Weight (g/mol) 86.84
PubChem CID 82050
ChEBI CHEBI:63042
Formula Weight 86.84
Concentration or Composition (by Analyte or Components) 4M exact strength on the certificate of analysis
Packaging AcroSeal™ Glass Bottle
Appearance Clear colorless to yellowish solution
Specific Gravity 1.18
Density 1.1800g/mL
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.