Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxypyridine 98.0+%, TCI America™
SDP

Catalog No. M169625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M169625G 25 g
M16965G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M169625G Supplier TCI America Supplier No. M169625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 620-08-6
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.128
InChI Key XQABVLBGNWBWIV-UHFFFAOYSA-N
Synonym pyridine, 4-methoxy, methyl pyridin-4-yl ether, 4-methoxy pyridine, .gamma.-methoxypyridine, 4-methoxypyridin, 4-methoxypyrdine, 4-methoxylpyridine, 4-methoxy-pyridine, 4-methoxypyridine, pubchem2585
PubChem CID 69278
IUPAC Name 4-methoxypyridine
SMILES COC1=CC=NC=C1

Specifications

Melting Point 4°C
Color Yellow
Boiling Point 190°C
Quantity 25 g
Formula Weight 109.13
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Methoxypyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.