Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxysalicylic acid, 99%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
2237-36-7
C8H8O4
168.15
MFCD00002450
MRIXVKKOHPQOFK-UHFFFAOYSA-N
4-methoxysalicylic acid,2-hydroxy-p-anisic acid,2-hydroxy-4-methoxy-benzoic acid,benzoic acid, 2-hydroxy-4-methoxy,2-hydroxy-4-methoxybenzoes,4-methoxysalicylicacid,pubchem18895,4-methoxy salicylic acid,4-methoxy-salicylic acid,acmc-1cs6b
75231
2-hydroxy-4-methoxybenzoic acid
COC1=CC(=C(C=C1)C(=O)O)O
2237-36-7
C8H8O4
168.15
MFCD00002450
MRIXVKKOHPQOFK-UHFFFAOYSA-N
4-methoxysalicylic acid,2-hydroxy-p-anisic acid,2-hydroxy-4-methoxy-benzoic acid,benzoic acid, 2-hydroxy-4-methoxy,2-hydroxy-4-methoxybenzoes,4-methoxysalicylicacid,pubchem18895,4-methoxy salicylic acid,4-methoxy-salicylic acid,acmc-1cs6b
75231
2-hydroxy-4-methoxybenzoic acid
COC1=CC(=C(C=C1)C(=O)O)O
Packaging Glass bottle
Melting Point 157.0°C to 159.0°C
Quantity 50g
Color White to Beige
Infrared Spectrum Authentic
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (HPLC)
Linear Formula CH3OC6H3(OH)CO2H
Beilstein 10, 379
Formula Weight 168.15
Physical Form Powder
Percent Purity 99%
Chemical Name or Material 4-Methoxysalicylic acid, 99%
Show More Show Less

EINECSNumber : 218-801-0