Learn More
4-Methoxystyrene, 98%, stab. with 0.1% 4-tert-butylcatechol
Description
4-Methoxystyrene is employed in the ferric chloride-catalyzed addition of activated methylenes to styrenes. It acts as a monomer in polymerization reactions. Further, it is used to prepare 1,1,2,2-Tetracyano-3-(p-methoxyphenyl)cyclobutane by reacting with ethenetetracarbonitrile.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4-Methoxystyrene is employed in the ferric chloride-catalyzed addition of activated methylenes to styrenes. It acts as a monomer in polymerization reactions. Further, it is used to prepare 1,1,2,2-Tetracyano-3-(p-methoxyphenyl)cyclobutane by reacting with ethenetetracarbonitrile.
Solubility
Slightly miscible with methanol.
Notes
Light sensitive. Store in cool place. Incompatible with strong oxidizing agents and strong acids.
Specifications
Specifications
| Density | 1.002 |
| Boiling Point | 204°C to 205°C |
| Flash Point | 72°C (161°F) |
| Refractive Index | 1.561 |
| Quantity | 5 g |
| Beilstein | 1098935 |
| Solubility Information | Slightly miscible with methanol. |
| Formula Weight | 134.18 |
| Percent Purity | 98% |
| Chemical Name or Material | 4-Methoxystyrene, stabilized with 0.1% 4-tert-butylcatechol |
Safety and Handling
| Hazard Category | H227-H315-H320 |
| Hazard Statement | GHS H Statement H227 Combustible liquid. |
| Precautionary Statement | P210-P233-P240-P241-P242-P243-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P374-P501c |
| EINECSNumber | 211-298-9 |
| TSCA | No |
| Recommended Storage | Keep cold |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.