Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxystyrene, 98%, stab. with 0.1% 4-tert-butylcatechol

Catalog No. p-7026380
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 637-69-4 | C9H10O | 134.18 g/mol

4-Methoxystyrene is employed in the ferric chloride-catalyzed addition of activated methylenes to styrenes. It acts as a monomer in polymerization reactions. Further, it is used to prepare 1,1,2,2-Tetracyano-3-(p-methoxyphenyl)cyclobutane by reacting with ethenetetracarbonitrile.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxystyrene is employed in the ferric chloride-catalyzed addition of activated methylenes to styrenes. It acts as a monomer in polymerization reactions. Further, it is used to prepare 1,1,2,2-Tetracyano-3-(p-methoxyphenyl)cyclobutane by reacting with ethenetetracarbonitrile.

Solubility
Slightly miscible with methanol.

Notes
Light sensitive. Store in cool place. Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 637-69-4
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.18
MDL Number MFCD00008619
InChI Key UAJRSHJHFRVGMG-UHFFFAOYSA-N
Synonym 4-methoxystyrene, 1-methoxy-4-vinylbenzene, p-methoxystyrene, 4-vinylanisole, benzene, 1-ethenyl-4-methoxy, p-vinylanisole, anisole, p-vinyl, unii-2ish8t4a6e, ccris 4381, 4-vinylanisol
PubChem CID 12507
IUPAC Name 1-ethenyl-4-methoxybenzene
SMILES COC1=CC=C(C=C)C=C1

Specifications

Density 1.002
Boiling Point 204°C to 205°C
Flash Point 72°C (161°F)
Refractive Index 1.561
Quantity 5 g
Beilstein 1098935
Solubility Information Slightly miscible with methanol.
Formula Weight 134.18
Percent Purity 98%
Chemical Name or Material 4-Methoxystyrene, stabilized with 0.1% 4-tert-butylcatechol
Hazard Category H227-H315-H320
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P374-P501c
EINECSNumber 211-298-9
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.