Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxystyrene, 96%, stabilized

Catalog No. p-158698
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle

CAS: 637-69-4 | C9H10O | 134.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 637-69-4
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.18
MDL Number MFCD00008619
InChI Key UAJRSHJHFRVGMG-UHFFFAOYSA-N
Synonym 4-methoxystyrene, 1-methoxy-4-vinylbenzene, p-methoxystyrene, 4-vinylanisole, benzene, 1-ethenyl-4-methoxy, p-vinylanisole, anisole, p-vinyl, unii-2ish8t4a6e, ccris 4381, 4-vinylanisol
PubChem CID 12507
IUPAC Name 1-ethenyl-4-methoxybenzene
SMILES COC1=CC=C(C=C)C=C1

Specifications

CAS Min % 95.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0000g/mL
Boiling Point 204°C to 205°C
Flash Point 76°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.5600 to 1.562
Linear Formula H2C=CHC6H4OCH3
Quantity 5 g
Beilstein 06, 561
Specific Gravity 1
Stabilizer 0.1% 4-tert-butylcatechol
Formula Weight 134.18
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 4-Methoxystyrene, 96%
Show More Show Less
EINECSNumber 211-298-9

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.