Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxythiobenzamide, 98%

Catalog No. aal1094503
Click to view available options
Quantity:
1 g
5 g

CAS: 2362-64-3 | C8H9NOS | 167.226 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2362-64-3
Molecular Formula C8H9NOS
Molecular Weight (g/mol) 167.226
MDL Number MFCD00040993
InChI Key WKWVTPKUHJOVTI-UHFFFAOYSA-N
Synonym 4-methoxythiobenzamide, 4-methoxybenzothioamide, 4-methoxybenzene-1-carbothioamide, benzenecarbothioamide, 4-methoxy, 4-methoxy-thiobenzamide, p-methoxythiobenzamide, amino 4-methoxyphenyl methane-1-thione, thio-p-anisamide, p-anisamide,thio, p-methoxybenzothiamide
PubChem CID 736827
IUPAC Name 4-methoxybenzenecarbothioamide
SMILES COC1=CC=C(C=C1)C(=S)N

Specifications

Melting Point 145°C to 147°C
Quantity 1 g
Beilstein 1072444
Formula Weight 167.23
Percent Purity 98%
Chemical Name or Material 4-Methoxythiobenzamide
Hazard Category H302+H332
Hazard Statement GHS H Statement
H301-H332
Toxic if swallowed.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.