Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methyl-1,3,2-dioxathiolane 2-Oxide (mixture of isomers) 98.0+%, TCI America™
SDP

Catalog No. M247125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M247125G 25 g
M24715G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M247125G Supplier TCI America Supplier No. M247125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1469-73-4
Molecular Formula C3H6O3S
Molecular Weight (g/mol) 122.138
MDL Number MFCD00041209
InChI Key SJHAYVFVKRXMKG-UHFFFAOYSA-N
Synonym 1,2-Propanediol Cyclic Sulfite, 1,2-Propyleneglycol Sulfite, 1,2-Propylene Sulfite
PubChem CID 15117
IUPAC Name 4-methyl-1,3,2-dioxathiolane 2-oxide
SMILES CC1COS(=O)O1

Specifications

Color Yellow
Boiling Point 40°C
Quantity 25 g
Formula Weight 122.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Methyl-1,3,2-dioxathiolane 2-Oxide (mixture of isomers)
RTECSNumber WT3500000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.