Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methyl-1,3-oxazol-2-amine, Thermo Scientific™

Catalog No. KM08202EA
Click to view available options
Quantity:
10 g

Chemical Identifiers

CAS 35629-70-0
Molecular Formula C4H6N2O
Molecular Weight (g/mol) 98.105
MDL Number MFCD00126682
InChI Key VCZJVXLWQTXSPQ-UHFFFAOYSA-N
PubChem CID 535824
IUPAC Name 4-methyl-1,3-oxazol-2-amine
SMILES CC1=COC(=N1)N

Specifications

Packaging Amber glass bottle
Quantity 10 g
Synonym 4-methyloxazol-2-amine, 2-amino-4-methyloxazole, 2-oxazolamine, 4-methyl, 4-methyl-oxazol-2-ylamine, 2-oxazolamine,4-methyl, 2-amino-4-methyl-1,3-oxazole, 2-amino-4-methyloxazol, 4-methyl-2-oxazolamine, 4-methyl-2-amino-oxazole, 2-amino-4-methyl-oxazole
Formula Weight 98.1
Percent Purity Tech.
Grade Technical
Chemical Name or Material 4-Methyl-1, 3-oxazol-2-amine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.