Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methyl-1-naphthoic Acid 98.0+%, TCI America™
SDP

Catalog No. M120325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M120325G 25 g
M12035G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M120325G Supplier TCI America Supplier No. M120325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4488-40-8
Molecular Formula C12H10O2
Molecular Weight (g/mol) 186.21
MDL Number MFCD00671557
InChI Key SIVYRLBDAPKADZ-UHFFFAOYSA-N
Synonym 4-methyl-1-naphthoic acid, 4-methyl-1-naphthalenecarboxylic acid, 4-methyl-1-napthoic acid, 1-naphthalenecarboxylic acid, 4-methyl, 4-methyl-naphthalene-1-carboxylic acid, 4-methylnaphthalenecarboxylic acid, 4-methylnaphthoic acid, acmc-1aehw, 4-methyl-1-naphthoicacid, 4-methyl-1-naphtholic acid
PubChem CID 3014285
IUPAC Name 4-methylnaphthalene-1-carboxylic acid
SMILES CC1=C2C=CC=CC2=C(C=C1)C(O)=O

Specifications

Melting Point 178°C
Color White-Yellow
Quantity 25 g
Formula Weight 186.21
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methyl-1-naphthoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.