Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methyl-1-pentene, 98+%

Catalog No. AA3302930
Click to view available options
Quantity:
10 g
50 g
250 g

CAS: 691-37-2 | C6H12 | 84.16 g/mol

4-Methyl-1-pentene is used as a monomer for olefin polymerisation. The resulting polymer is poly(4-methyl-1-pentene). It can also be used to produce 1,2-diiodo-4-methyl-pentane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 691-37-2
Molecular Formula C6H12
Molecular Weight (g/mol) 84.16
MDL Number MFCD00008949
InChI Key WSSSPWUEQFSQQG-UHFFFAOYSA-N
Synonym 4-methyl-1-pentene, 1-pentene, 4-methyl, poly 4-methyl-1-pentene, unii-x10hrj2y7w, x10hrj2y7w, poly 4-methylpentene, isobutylethene, isohexene, 4-methylpentene-1, 1-pentene, 4-methyl-, homopolymer
PubChem CID 12724
IUPAC Name 4-methylpent-1-ene
SMILES CC(C)CC=C

Specifications

Melting Point -153°C
Color Colorless
Density 0.664 g/mL
Boiling Point 53°C to 54°C
Flash Point -32°C (-25°F)
Odor Petrol-like
Refractive Index 1.382
Linear Formula (CH3)2CHCH2CH=CH2
Quantity 250 g
UN Number UN3295
Beilstein 1731096
Solubility Information Soluble in alcohol,benzene,chloroform,petroleum ether. Insoluble in water.
Formula Weight 84.16
Percent Purity ≥98%
Physical Form Liquid
Chemical Name or Material 4-Methyl-1-pentene
Show More Show Less
Hazard Category H225
Hazard Statement GHS H Statement
H225-H304
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber 211-720-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.