Learn More
4-Methyl-1-pentene, 98+%
Description
4-Methyl-1-pentene is used as a monomer for olefin polymerisation. The resulting polymer is poly(4-methyl-1-pentene). It can also be used to produce 1,2-diiodo-4-methyl-pentane.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | -153°C |
| Color | Colorless |
| Density | 0.664 g/mL |
| Boiling Point | 53°C to 54°C |
| Flash Point | -32°C (-25°F) |
| Odor | Petrol-like |
| Refractive Index | 1.382 |
| Linear Formula | (CH3)2CHCH2CH=CH2 |
| Quantity | 10 g |
| UN Number | UN3295 |
| Show More |
Safety and Handling
| Hazard Category | H225 |
| Hazard Statement | GHS H Statement H225-H304 Highly flammable liquid and vapor. May be fatal if swallowed and enters airways. |
| Precautionary Statement | P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S. |
| EINECSNumber | 211-720-1 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.