Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methyl-2-hexylamine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. M24091G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M24091G 1 g
M24095G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M24091G Supplier TCI America Supplier No. M24091G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13803-74-2
Molecular Formula C7H18ClN
Molecular Weight (g/mol) 151.678
MDL Number MFCD09038354
InChI Key ZKKBPHUAHARETG-UHFFFAOYSA-N
Synonym Methylhexanamine Hydrochloride, 1,3-Dimethylpentylamine Hydrochloride, 1,3-Dimethylamylamine Hydrochloride, 2-Amino-4-methylhexane Hydrochloride
PubChem CID 44118858
IUPAC Name 4-methylhexan-2-amine;hydrochloride
SMILES CCC(C)CC(C)N.Cl

Specifications

Melting Point 130°C
Color White-Yellow
Quantity 1 g
Formula Weight 151.68
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methyl-2-hexylamine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.