Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methyl-3-nitrobenzaldehyde, 98%

Catalog No. aal1345403
Click to view available options
Quantity:
1 g
5 g
25 g

CAS: 31680-07-6 | C8H7NO3 | 165.15 g/mol

4-Methyl-3-nitrobenzaldehyde is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methyl-3-nitrobenzaldehyde is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents. It is sensitive to air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31680-07-6
Molecular Formula C8H7NO3
Molecular Weight (g/mol) 165.15
MDL Number MFCD00017011
InChI Key KHWGAWBXQOKXIJ-UHFFFAOYSA-N
Synonym 3-nitro-p-tolualdehyde, 4-methyl-3-nitro-benzaldehyde, 3-nitro-4-methylbenzaldehyde, benzaldehyde, 4-methyl-3-nitro, methylnitrobenzaldehyde, pubchem8157, acmc-209hod, 4-methyl-3-nitrobenzaldehyde
PubChem CID 1798998
IUPAC Name 4-methyl-3-nitrobenzaldehyde
SMILES CC1=CC=C(C=O)C=C1[N+]([O-])=O

Specifications

Melting Point 44°C to 48°C
Flash Point >110°C (230°F)
Quantity 1 g
Beilstein 1637706
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 165.15
Percent Purity 98%
Chemical Name or Material 4-Methyl-3-nitrobenzaldehyde
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 250-760-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.