Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methyl-3-nitrobenzoic acid, 99%

Catalog No. p-165175
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle

CAS: 96-98-0 | C8H7NO4 | 181.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-98-0
Molecular Formula C8H7NO4
Molecular Weight (g/mol) 181.15
MDL Number MFCD00007174
InChI Key BBEWSMNRCUXQRF-UHFFFAOYSA-N
PubChem CID 7319
SMILES CC1=C(C=C(C=C1)C(=O)O)[N+](=O)[O-]

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 187.0°C to 190.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3C6H3(NO2)CO2H
Quantity 25 g
Beilstein 09, 502
Synonym 4-methyl-3-nitrobenzoic acid, 3-nitro-4-methylbenzoic acid, 3-nitro-p-toluic acid, benzoic acid, 4-methyl-3-nitro, 3-nitro-para-toluic acid, m-nitro-p-toluic acid, 4-methyl-3-nitro-benzoic acid, p-toluic acid, 3-nitro, 3-nitro-4-methyl benzoic acid, benzoic acid,4-methyl-3-nitro
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohols
Formula Weight 181.15
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 4-Methyl-3-nitrobenzoic acid, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
EINECSNumber 202-549-3
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.