Alfa Aesar™ 4-Methyl-3-nitrophenol, 98%

Manufacturer: Alfa Aesar™ 4428509

Quantity 10g

 View more versions of this product

Catalog No. AA4428509

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2042-14-0
    Molecular Formula C7H7NO3
    Formula Weight 153.14
    MDL Number MFCD00007244
    UN Number UN2446
    Synonym 3-nitro-p-cresol
    Chemical Name or Material 4-Methyl-3-nitrophenol

    Specifications

    Formula Weight 153.14
    Health Hazard 3 GHS P Statement
    P280h-P305+P351+P338-P309-P310
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Synonym 3-nitro-p-cresol
    Molecular Formula C7H7NO3
    CAS 2042-14-0
    UN Number UN2446
    Physical Form Powder
    Melting Point 77° to 81°C
    Quantity 10g
    Odor Phenol-like
    Health Hazard 2 GHS H Statement
    H311-H302-H332-H315-H319-H335
    Toxic in contact with skin.
    Harmful if swallowed.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    MDL Number MFCD00007244
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITROCRESOLS, SOLID
    Chemical Name or Material 4-Methyl-3-nitrophenol
    Assay 98%

    GHS H Statement
    H311-H302-H332-H315-H319-H335
    Toxic in contact with skin.
    Harmful if swallowed.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280h-P305+P351+P338-P309-P310
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit