Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4-Methyl-3-nitrophenylboronic Acid (contains varying amounts of Anhydride), TCI America™
Shop All Tokyo Chemical Industry (TCI) Products

Click to view available options
Quantity:
1 g
5 g
Specifications
Specifications
| Chemical Name or Material | 4-Methyl-3-nitrophenylboronic Acid (contains varying amounts of Anhydride) |
| CAS | 80500-27-2 |
| Molecular Formula | C7H8BNO4 |
| Quantity | 1 g |
| MDL Number | MFCD00191550 |
| Synonym | 4-methyl-3-nitrophenyl boronic acid, 4-methyl-3-nitrobenzeneboronic acid, 3-nitro-p-tolylboronic acid, 4-methyl-3-nitrophenyl boranediol, 4-methyl-3-nitro-phenyl boronic acid, 3-nitro-4-methylphenylboronic acid, 2-nitrotoluene-4-boronic acid, boronic acid, 4-methyl-3-nitrophenyl, 3-nitro-4-methylbenzeneboronic acid |
| InChI Key | OASVXBRTNVFKFS-UHFFFAOYSA-N |
| SMILES | B(C1=CC(=C(C=C1)C)[N+](=O)[O-])(O)O |
| IUPAC Name | (4-methyl-3-nitrophenyl)boronic acid |
| Molecular Weight (g/mol) | 180.954 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction