Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methyl-3-nitrothiobenzamide, 97%, Thermo Scientific™

Catalog No. AAH5470906
Click to view available options
Quantity:
250 mg
1 g
5 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1233513-42-2
Molecular Formula C8H8N2O2S
Molecular Weight (g/mol) 196.22
MDL Number MFCD09025715
InChI Key PGQMEJDSPAENNP-UHFFFAOYSA-N
Synonym 4-methyl-3-nitrothiobenzamide
PubChem CID 45040726
IUPAC Name 4-methyl-3-nitrobenzenecarbothioamide
SMILES CC1=C(C=C(C=C1)C(N)=S)[N+]([O-])=O

Specifications

Quantity 5 g
Formula Weight 196.22
Percent Purity 97%
Chemical Name or Material 4-Methyl-3-nitrothiobenzamide
Hazard Category Danger
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.