Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methyl-5-thiazoleethanol, 98%, ACROS Organics™

Click to view available options
Quantity:
25mL
100mL
Packaging:
Glass bottle
137-00-8
C6H9NOS
143.21
MFCD00005339
BKAWJIRCKVUVED-UHFFFAOYSA-N
4-methyl-5-thiazoleethanol,sulfurol,5-2-hydroxyethyl-4-methylthiazole,hemineurine,2-4-methylthiazol-5-yl ethanol,4-methyl-5-thiazolethanol,5-thiazoleethanol, 4-methyl,2-4-methyl-1,3-thiazol-5-yl ethanol,4-methyl-5-hydroxyethylthiazole,2-4-methyl-1,3-thiazol-5-yl ethan-1-ol
1136
CHEBI:17957
2-(4-methyl-1,3-thiazol-5-yl)ethanol
CC1=C(SC=N1)CCO
137-00-8
C6H9NOS
143.21
MFCD00005339
BKAWJIRCKVUVED-UHFFFAOYSA-N
4-methyl-5-thiazoleethanol,sulfurol,5-2-hydroxyethyl-4-methylthiazole,hemineurine,2-4-methylthiazol-5-yl ethanol,4-methyl-5-thiazolethanol,5-thiazoleethanol, 4-methyl,2-4-methyl-1,3-thiazol-5-yl ethanol,4-methyl-5-hydroxyethylthiazole,2-4-methyl-1,3-thiazol-5-yl ethan-1-ol
1136
CHEBI:17957
2-(4-methyl-1,3-thiazol-5-yl)ethanol
CC1=C(SC=N1)CCO
Density 1.1900g/mL
Refractive Index 1.5480 to 1.551
Boiling Point 135°C (7.0mmHg)
Flash Point >110°C
Specific Gravity 1.19
Infrared Spectrum Authentic
Quantity 100mL
Packaging Glass bottle
Water 1% max.
Color Yellow to Brown
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Beilstein 27, 1754
Merck Index 15, 6199
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in alcohol,ether,benzene and chloroform
Formula Weight 143.21
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 4-Methyl-5-thiazoleethanol, 98%
Show More Show Less

GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning

EINECSNumber : 205-272-6

TSCA : TSCA