Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methyl(thiobenzamide), 97%

Catalog No. p-7025019
Click to view available options
Quantity:
1 g
5 g

CAS: 2362-62-1 | C8H9NS | 151.227 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2362-62-1
Molecular Formula C8H9NS
Molecular Weight (g/mol) 151.227
MDL Number MFCD00173750
InChI Key QXYZSNGZMDVLKN-UHFFFAOYSA-N
Synonym 4-methylbenzothioamide, 4-methyl thiobenzamide, 4-methylthiobenzamide, 4-methylbenzene-1-carbothioamide, 4-methyl-thiobenzamide, 4-methylbenzenethioamide, benzenecarbothioamide, 4-methyl, amino 4-methylphenyl methane-1-thione, 4-pmtb, p-methylthiobenzoylamide
PubChem CID 737209
IUPAC Name 4-methylbenzenecarbothioamide
SMILES CC1=CC=C(C=C1)C(=S)N

Specifications

Melting Point 166°C to 169°C
Quantity 1 g
Formula Weight 151.23
Percent Purity 97%
Chemical Name or Material 4-Methyl(thiobenzamide)
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.