Learn More
4'-Methylacetophenone, 96%
Description
4'-Methylacetophenone is used as a flavoring agent. It reacts with morpholine to get 4-(p-tolyl-thioacetyl)-morpholine in the presence of sulfur as a reagent. Further, it is used as an intermediate in the manufacture of active pharmaceutical ingredients, perfumes and cosmetics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4′-Methylacetophenone is used as a flavoring agent. It reacts with morpholine to get 4-(p-tolyl-thioacetyl)-morpholine in the presence of sulfur as a reagent. Further, it is used as an intermediate in the manufacture of active pharmaceutical ingredients, perfumes and cosmetics.
Solubility
Miscible with water.
Notes
Incompatible with strong oxidizing agents, strong bases and strong reducing agents.

Specifications
Specifications
Melting Point | -19°C |
Density | 1.01 |
Boiling Point | 224°C to 226°C |
Flash Point | 92°C (197°F) |
Refractive Index | 1.534 |
Quantity | 1000 g |
Beilstein | 606053 |
Solubility Information | Miscible with water. |
Formula Weight | 134.18 |
Percent Purity | 96% |
Show More |
Safety and Handling
Hazard Category | H227-H302-H315-H319 |
Hazard Statement | GHS H Statement H301-H227 Toxic if swallowed. Combustible liquid. |
Precautionary Statement | P210-P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P370+P378q-P501c |
EINECSNumber | 204-514-8 |
RTECSNumber | AM9463000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.