Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methylaminophenol sulfate, 99%, extra pure

Catalog No. p-159619
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC156800050 5 g
AC156801000 100 g
AC156805000 500 g
AC156800025 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC156800050 Supplier Thermo Scientific Chemicals Supplier No. 156800050
Only null left
Add to Cart

4-Methylaminophenol sulfate, 99%, C14H18N2O2.H2SO4, CAS Number-55-55-0, Metol, 500g, 13, 441, 100.0, 98.5, Beige to White or Gray, Decomposition Information: >260 deg.C, 1.5770g/mL, 200-237-1, 344.38, Extra Pure, GHS Signal Word: Warning | CAS: 55-55-0 | C14H18N2O2·H2SO4 | 344.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55-55-0
Molecular Formula C14H18N2O2·H2SO4
Molecular Weight (g/mol) 344.38
MDL Number MFCD00013140
Synonym Metol

Specifications

CAS Min % 98.5
CAS Max % 100.0
Decomposition Information Decomposition Information: >260°C
Melting Point 260.0°C
Color Beige to White or Gray
Density 1.5770g/mL
Ignition Residue 0.15% max.
Loss on Drying 0.3% max.
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Linear Formula (CH3NHC6H4OH)2·H2SO4
Quantity 5 g
Beilstein 13, 441
Specific Gravity 1.577
Merck Index 15, 6091
Solubility Information Solubility in water: 50g/L (20°C). Other solubilities: soluble in 20 parts cold water,soluble in 6 parts boiling water,slightly soluble in alcohol,insoluble in ether
Formula Weight 344.38
Percent Purity 99%
Grade Extra Pure
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 4-Methylaminophenol sulfate, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Very toxic to aquatic life with long lasting effects.
May cause an allergic skin reaction.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Precautionary Statement Emergency Overview
Harmful if swallowed. May cause eye, skin, and respiratory tract irritation . May cause an allergic skin reaction. Danger of serious damage to health by prolonged exposure if swallowed. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment.
NFPA
Health: 2
Flammability: 1
Instability: 1
EINECSNumber 200-237-1
RTECSNumber SL8650000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.