Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methylbenzamidoxime, 97%, Thermo Scientific™

Catalog No. p-7034056
Click to view available options
Quantity:
1 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19227-13-5
Molecular Formula C8H10N2O
Molecular Weight (g/mol) 150.181
MDL Number MFCD00019952
InChI Key NKJXMLIWSJATEE-UHFFFAOYSA-N
Synonym 4-methylbenzamidoxime, 4-methylbenzamide oxime, p-toluamidoxime, n-hydroxy-4-methyl-benzamidine, hydroxyimino 4-methylphenyl methylamine, n'-hydroxy-4-methylbenzene-1-carboximidamide, e-n'-hydroxy-4-methylbenzenecarboximidamide, z-n'-hydroxy-4-methylbenzene-1-carboximidamide, n'-hydroxy-4-methylbenzimidamide, n-hydroxy-4-methylbenzenecarboximidamide
PubChem CID 5493168
IUPAC Name N'-hydroxy-4-methylbenzenecarboximidamide
SMILES CC1=CC=C(C=C1)C(=NO)N

Specifications

Melting Point 135°C to 138°C
Quantity 1 g
Formula Weight 150.18
Percent Purity 97%
Chemical Name or Material 4-Methylbenzamidoxime
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
RTECSNumber XS4870000
TSCA No
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.