Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methylbenzhydrylamine hydrochloride, polymer-supported, 1% cross-linked, 100-200 mesh, 0.5-1.0 mmol/g on PS-DVB

Catalog No. p-7059351
Click to view available options
Quantity:
5 g
25 g

CAS: 5267-49-2 | C14H16ClN | 233.74 g/mol

Hydrochloride resins have been used to study the treatment of hyperphosphataemia in patients with end-stage renal failure. And also used for attachment of linkers in a similar manner to PL-AMS Resin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5267-49-2
Molecular Formula C14H16ClN
Molecular Weight (g/mol) 233.74
MDL Number MFCD00801150
InChI Key NHTZSJKMWBONMD-UHFFFAOYNA-N
Synonym 4-methylphenyl phenyl methanamine hydrochloride, 4-methylphenyl phenyl methyl amine hydrochloride, 4-methylbenzhydrylamine hydrochloride, phenyl p-tolyl methanamine hydrochloride, 4-methylphenyl phenyl methyl amine hcl, 4-methylphenyl-phenylmethanamine hydrochloride
PubChem CID 2734294

Specifications

Color White to Yellow
Mesh Size 100 to 200 Mesh
Quantity 5 g
Solubility Information Suspended in DCM/DFM(1:1).
Physical Form Beads
Chemical Name or Material 4-Methylbenzhydrylamine hydrochloride, Polymer-supported, 1% cross-linked, 0.5 to 1.0 mmol/g on PS-DVB
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.