Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methylbenzohydrazide 98.0+%, TCI America™
SDP

Catalog No. p-7133470
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 3619-22-5
Molecular Formula C8H10N2O
Molecular Weight (g/mol) 150.18
MDL Number MFCD00007607
InChI Key MFFVZXOPRXMVET-UHFFFAOYSA-N
Synonym p-toluic hydrazide, p-toluic acid hydrazide, p-toluoylhydrazine, 4-methylbenzoylhydrazine, p-toluic acid, hydrazide, 4-methylbenzoic acid hydrazide, p-methylbenzohydrazide, p-toluohydrazide, p-methylbenzoylhydrazine, 4-toluic acid hydrazide
PubChem CID 77174
IUPAC Name 4-methylbenzohydrazide
SMILES CC1=CC=C(C=C1)C(=O)NN

Specifications

Melting Point 116°C
Color White-Yellow
Quantity 5 g
Formula Weight 150.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methylbenzohydrazide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.