Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methylbenzylmagnesium chloride, 0.5M solution in THF, AcroSeal™

Catalog No. AC433700500
Change view
Click to view available options
Quantity:
50 mL
Packaging:
AcroSeal™ Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC433700500 50 mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC433700500 Supplier Thermo Scientific Chemicals Supplier No. 433700500
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 29875-07-8 | C8H9ClMg | 164.92 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29875-07-8 , 109-99-9
Molecular Formula C8H9ClMg
Molecular Weight (g/mol) 164.92
MDL Number MFCD01319893
InChI Key SAKYCLWKOWGTFT-UHFFFAOYSA-M
Synonym 4-methylbenzylmagnesium chloride, 4-methylbenzylmagnesium chloride solution, p-methylbenzylmagnesium chloride, 4-methylbenzyl magnesium chloride, sakyclwkowgtft-uhfffaoysa-m, magnesium,chloro 4-methylphenyl methyl, chloro 4-methylphenyl methyl magnesium, 4-methylbenzylmagnesium chloride solution, 0.5 m in thf, 4-methylbenzylmagnesium chloride 0.5 m in tetrahydrofuran
PubChem CID 11298156
IUPAC Name magnesium;1-methanidyl-4-methylbenzene;chloride
SMILES CC1=CC=C(C[Mg]Cl)C=C1

Specifications

CAS Min % 8.08, 90.12
CAS Max % 9.88, 91.92
Color Brown or Green to Yellow
Density 0.9180g/mL
Boiling Point 65.0°C
Flash Point −17°C
Packaging AcroSeal™ Glass Bottle
Quantity 50 mL
Specific Gravity 0.918
Solubility Information Solubility in water: reacts
Formula Weight 164.92
Physical Form Solution
Chemical Name or Material 4-Methylbenzylmagnesium chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
Reacts violently with water.
May form explosive peroxides.
Suspected of causing cancer.<br/
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.